Product Details

SNP ID
rs121917747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:72891502 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTTACTGGAAAAGGACGAGTTC[A/T]AGTCTGGAGCCCACGTGGACTTCTA
Phenotype
MIM: 182125
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SPR PubMed Links

Gene Details

Gene
SPR
Gene Name
sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003124.4 801 Nonsense Mutation AAG,TAG K251* NP_003115.1

View Full Product Details