Product Details

SNP ID
rs137862503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241799073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGCCCTTTCCACAGATACTTCC[A/G]GGTCATCCTCCTCCTCCTCCTGGCC
Phenotype
MIM: 608237 MIM: 608527
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAL3ST2 PubMed Links

Gene Details

Gene
GAL3ST2
Gene Name
galactose-3-O-sulfotransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022134.2 169 Missense Mutation CAG,CGG Q13R NP_071417.2
Gene
NEU4
Gene Name
neuraminidase 4
There are no transcripts associated with this gene.

View Full Product Details