Product Details

SNP ID
rs137998852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:173910287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGAAAAAAATTAAGTTACTTGGT[A/T]TTAAAAATTCAACTTTAATAGCTCA
Phenotype
MIM: 601804
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SP3 PubMed Links

Gene Details

Gene
SP3
Gene Name
Sp3 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017371.4 Intron NP_001017371.3
NM_001172712.1 Intron NP_001166183.1
NM_003111.4 Intron NP_003102.1

View Full Product Details