Product Details

SNP ID
rs138045213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:10441703 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCCATATGCTGGATGAATAATA[C/G]TTCTCATCTGGTTTAGGTCTCTATA
Phenotype
MIM: 165640
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929715 PubMed Links

Gene Details

Gene
LOC101929715
Gene Name
uncharacterized LOC101929715
There are no transcripts associated with this gene.

Gene
ODC1
Gene Name
ornithine decarboxylase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287188.1 1608 Missense Mutation AAC,AAG N220K NP_001274117.1
NM_001287189.1 1608 Missense Mutation AAC,AAG N349K NP_001274118.1
NM_001287190.1 1608 Missense Mutation AAC,AAG N349K NP_001274119.1
NM_002539.2 1608 Missense Mutation AAC,AAG N349K NP_002530.1
Gene
SNORA80B
Gene Name
small nucleolar RNA, H/ACA box 80B
There are no transcripts associated with this gene.

View Full Product Details