Product Details

SNP ID
rs138056768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240042962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGAAGGGCCGTGGATGAAGAGC[C/T]GTGGATGAACAGGCATGCATGTGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR6B3 PubMed Links

Gene Details

Gene
OR6B3
Gene Name
olfactory receptor family 6 subfamily B member 3
There are no transcripts associated with this gene.

Gene
PRR21
Gene Name
proline rich 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080835.1 21 Silent Mutation ACA,ACG T7T NP_001074304.1

View Full Product Details