Product Details

SNP ID
rs138076878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:79085530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCTTGCACAGTAGCCAGCATTA[A/G]CACTGCTCGGGGATCCAGTGTCCCA
Phenotype
MIM: 167771
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
REG1B PubMed Links

Gene Details

Gene
REG1B
Gene Name
regenerating family member 1 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006507.3 507 Missense Mutation NP_006498.1

View Full Product Details