Product Details
- SNP ID
-
rs138164068
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:158172148 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATACAGTAGACTCCATGTCCTTTC[C/T]TGGAGGTTTTTGAGGACTAATTTTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC148
PubMed Links
Gene Details
- Gene
- CCDC148
- Gene Name
- coiled-coil domain containing 148
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001301684.1 |
1990 |
Missense Mutation |
AGA,GGA |
R435G |
NP_001288613.1 |
NM_001301685.1 |
1990 |
Intron |
|
|
NP_001288614.1 |
NM_138803.3 |
1990 |
Missense Mutation |
AGA,GGA |
R581G |
NP_620158.3 |
XM_005246319.3 |
1990 |
Missense Mutation |
AGA,GGA |
R602G |
XP_005246376.1 |
XM_011510638.2 |
1990 |
Missense Mutation |
AGA,GGA |
R615G |
XP_011508940.1 |
XM_017003392.1 |
1990 |
Missense Mutation |
AGA,GGA |
R594G |
XP_016858881.1 |
XM_017003393.1 |
1990 |
Missense Mutation |
AGA,GGA |
R580G |
XP_016858882.1 |
XM_017003394.1 |
1990 |
Missense Mutation |
AGA,GGA |
R541G |
XP_016858883.1 |
XM_017003395.1 |
1990 |
Missense Mutation |
AGA,GGA |
R508G |
XP_016858884.1 |
XM_017003396.1 |
1990 |
Missense Mutation |
AGA,GGA |
R495G |
XP_016858885.1 |
XM_017003397.1 |
1990 |
Missense Mutation |
AGA,GGA |
R435G |
XP_016858886.1 |
XM_017003398.1 |
1990 |
Missense Mutation |
AGA,GGA |
R368G |
XP_016858887.1 |
- Gene
- CCDC148-AS1
- Gene Name
- CCDC148 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details