Product Details

SNP ID
rs138231883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112485985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGACAAACTGTGTCGCAAAGCT[G/T]CTTTAGTGAAGTAAGTGTTAAGACC
Phenotype
MIM: 608291
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TTL PubMed Links

Gene Details

Gene
TTL
Gene Name
tubulin tyrosine ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153712.4 444 Missense Mutation GCT,TCT A76S NP_714923.1
XM_005263599.3 444 Missense Mutation GCT,TCT A76S XP_005263656.1
XM_011510665.2 444 Missense Mutation GCT,TCT A76S XP_011508967.1

View Full Product Details