Product Details

SNP ID
rs138249576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:31337672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAACTTGTCCACGCAGGCATTG[C/T]GGATCTTCTCCGGGGTGGCAGGGCT
Phenotype
MIM: 607633
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
XDH PubMed Links

Gene Details

Gene
XDH
Gene Name
xanthine dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000379.3 3999 Missense Mutation NP_000370.2
XM_011533095.2 3999 Missense Mutation XP_011531397.1
XM_011533096.2 3999 Intron XP_011531398.1

View Full Product Details