Product Details

SNP ID
rs138290494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:26310231 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGGCAACCAAACAATAGGATGA[A/G]GACGCCCTGAGAAGAGGGACATGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRF3 PubMed Links

Gene Details

Gene
ADGRF3
Gene Name
adhesion G protein-coupled receptor F3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145168.1 2889 Missense Mutation CTC,TTC L1015F NP_001138640.1
NM_001145169.1 2889 Missense Mutation CTC,TTC L946F NP_001138641.1
NM_001321971.1 2889 Missense Mutation CTC,TTC L947F NP_001308900.1
NM_001321975.1 2889 Missense Mutation CTC,TTC L618F NP_001308904.1
NM_153835.3 2889 Missense Mutation CTC,TTC L816F NP_722577.2
XM_011532621.2 2889 Missense Mutation CTC,TTC L956F XP_011530923.1
XM_011532622.2 2889 Missense Mutation CTC,TTC L946F XP_011530924.1
XM_011532623.2 2889 Missense Mutation CTC,TTC L940F XP_011530925.1
XM_017003505.1 2889 Missense Mutation CTC,TTC L945F XP_016858994.1
XM_017003506.1 2889 Missense Mutation CTC,TTC L769F XP_016858995.1

View Full Product Details