Product Details
- SNP ID
-
rs138459633
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:182834856 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTTTGTATTTCGGGATTTAGTTG[C/T]GTGCTTGCCGGGGGTTCGAGTTCCT
- Phenotype
-
MIM: 605083
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FRZB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7775] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FRZB
- Gene Name
- frizzled-related protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001463.3 |
1189 |
Missense Mutation |
CAC,CGC |
H324R |
NP_001454.2 |
View Full Product Details