Product Details

SNP ID
rs138621625
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:132674799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTATGGGAATATTTGAAAATCATC[A/C]TAGATCTAGTGCTTATTTCATTATA
Phenotype
MIM: 610450 MIM: 608789
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
LYPD1 PubMed Links

Gene Details

Gene
LYPD1
Gene Name
LY6/PLAUR domain containing 1
There are no transcripts associated with this gene.

Gene
NCKAP5
Gene Name
NCK associated protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207363.2 Intron NP_997246.2
NM_207481.3 Intron NP_997364.3
XM_005263659.3 Intron XP_005263716.1
XM_005263660.3 Intron XP_005263717.1
XM_011511097.2 Intron XP_011509399.1
XM_011511098.2 Intron XP_011509400.1
XM_011511099.2 Intron XP_011509401.1
XM_011511100.2 Intron XP_011509402.1
XM_011511101.1 Intron XP_011509403.1
XM_011511102.2 Intron XP_011509404.1
XM_011511103.2 Intron XP_011509405.1
XM_011511104.1 Intron XP_011509406.1
XM_011511105.2 Intron XP_011509407.1
XM_017003974.1 Intron XP_016859463.1
XM_017003975.1 Intron XP_016859464.1
XM_017003976.1 Intron XP_016859465.1
XM_017003977.1 Intron XP_016859466.1
XM_017003978.1 Intron XP_016859467.1
XM_017003979.1 Intron XP_016859468.1
XM_017003980.1 Intron XP_016859469.1

View Full Product Details