Product Details
- SNP ID
-
rs138709525
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:26310941 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTATGATGGCCAGCACTGGCCCCAC[A/G]AAGGTGTATAACGCCCCTCCCTTCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ADGRF3
PubMed Links
Gene Details
- Gene
- ADGRF3
- Gene Name
- adhesion G protein-coupled receptor F3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001145168.1 |
2633 |
Silent Mutation |
TTC,TTT |
F929F |
NP_001138640.1 |
NM_001145169.1 |
2633 |
Silent Mutation |
TTC,TTT |
F860F |
NP_001138641.1 |
NM_001321971.1 |
2633 |
Silent Mutation |
TTC,TTT |
F861F |
NP_001308900.1 |
NM_001321975.1 |
2633 |
Silent Mutation |
TTC,TTT |
F532F |
NP_001308904.1 |
NM_153835.3 |
2633 |
Silent Mutation |
TTC,TTT |
F730F |
NP_722577.2 |
XM_011532621.2 |
2633 |
Silent Mutation |
TTC,TTT |
F870F |
XP_011530923.1 |
XM_011532622.2 |
2633 |
Silent Mutation |
TTC,TTT |
F860F |
XP_011530924.1 |
XM_011532623.2 |
2633 |
Silent Mutation |
TTC,TTT |
F854F |
XP_011530925.1 |
XM_017003505.1 |
2633 |
Silent Mutation |
TTC,TTT |
F859F |
XP_016858994.1 |
XM_017003506.1 |
2633 |
Silent Mutation |
TTC,TTT |
F683F |
XP_016858995.1 |
View Full Product Details