Product Details

SNP ID
rs138752223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74527747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGACCACATACCTGACTGGGGG[C/T]GCAATCTGGGGTGTCTTTGTCGCTT
Phenotype
MIM: 602434 MIM: 606441 MIM: 607163
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AUP1 PubMed Links

Gene Details

Gene
AUP1
Gene Name
ancient ubiquitous protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181575.4 1043 Missense Mutation CAC,CGC H277R NP_853553.1
XM_005264392.3 1043 Missense Mutation CAC,CGC H334R XP_005264449.1
Gene
DQX1
Gene Name
DEAQ-box RNA dependent ATPase 1
There are no transcripts associated with this gene.

Gene
HTRA2
Gene Name
HtrA serine peptidase 2
There are no transcripts associated with this gene.

Gene
LOXL3
Gene Name
lysyl oxidase like 3
There are no transcripts associated with this gene.

View Full Product Details