Product Details

SNP ID
rs138915136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240126620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCCCTTCTTCTCCACCAGCAAC[A/G]GATGGTGTTTCATCACTCTTAAAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPS9 PubMed Links

Gene Details

Gene
COPS9
Gene Name
COP9 signalosome subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163424.1 1127 Intron NP_001156896.1
NM_138336.1 1127 Silent Mutation NP_612209.1
XM_017003411.1 1127 Silent Mutation XP_016858900.1

View Full Product Details