Product Details
- SNP ID
-
rs139153499
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:68974667 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCTACTCCTCTGTCCACTGCTTTC[A/G]TGAAGACAAGATGAAGTTCACAGTG
- Phenotype
-
MIM: 606402
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GKN1
PubMed Links
Gene Details
- Gene
- GKN1
- Gene Name
- gastrokine 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019617.3 |
95 |
Missense Mutation |
CAT,CGT |
H11R |
NP_062563.3 |
View Full Product Details