Product Details

SNP ID
rs139157386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233060598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCGAGTCCATCTCCCGGGCATA[C/T]GCGCTCTGCGTGCTGTGAGTACAAC
Phenotype
MIM: 601582
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INPP5D PubMed Links

Gene Details

Gene
INPP5D
Gene Name
inositol polyphosphate-5-phosphatase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017915.2 632 Silent Mutation TAC,TAT Y40Y NP_001017915.1
NM_005541.4 632 Silent Mutation TAC,TAT Y40Y NP_005532.2
XM_017004004.1 632 UTR 5 XP_016859493.1

View Full Product Details