Product Details

SNP ID
rs139240350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:10572126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTTCAGGTGTCCGGCCGAACGA[C/T]GGAGTCTTTTCCTTTCTTGTCGATG
Phenotype
MIM: 616197
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOL10 PubMed Links

Gene Details

Gene
NOL10
Gene Name
nucleolar protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261392.1 1663 Missense Mutation CAT,CGT H645R NP_001248321.1
NM_001261394.1 1663 Missense Mutation CAT,CGT H621R NP_001248323.1
NM_024894.3 1663 Missense Mutation CAT,CGT H671R NP_079170.2
XM_011510398.2 1663 Missense Mutation CAT,CGT H521R XP_011508700.1
XM_011510399.2 1663 Missense Mutation CAT,CGT H521R XP_011508701.1
XM_011510400.2 1663 Intron XP_011508702.1
XM_011510402.2 1663 Intron XP_011508704.1
XM_017004990.1 1663 Missense Mutation CAT,CGT H346R XP_016860479.1
XM_017004991.1 1663 Missense Mutation CAT,CGT H346R XP_016860480.1

View Full Product Details