Product Details

SNP ID
rs139416675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:71134093 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAAGAAGAAGCAGAAGAACTAA[A/G]TATTTCGGAAACGTGAGTATTTTGA
Phenotype
MIM: 608419 MIM: 605503
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MCEE PubMed Links

Gene Details

Gene
MCEE
Gene Name
methylmalonyl-CoA epimerase
There are no transcripts associated with this gene.

Gene
MPHOSPH10
Gene Name
M-phase phosphoprotein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005791.2 1266 Missense Mutation AAT,AGT N305S NP_005782.1

View Full Product Details