Product Details

SNP ID
rs139443908
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:203436032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAGCTCCTAATAACCTTGATGTC[A/T]GAGTTGTCACTTGGTTGGAATTATT
Phenotype
MIM: 606442 MIM: 609035
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ABI2 PubMed Links

Gene Details

Gene
ABI2
Gene Name
abl-interactor 2
There are no transcripts associated with this gene.

Gene
RAPH1
Gene Name
Ras association (RalGDS/AF-6) and pleckstrin homology domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203365.3 7239 Intron NP_976241.1
NM_213589.2 7239 UTR 3 NP_998754.1
XM_005246777.3 7239 UTR 3 XP_005246834.1
XM_005246779.4 7239 UTR 3 XP_005246836.1
XM_006712695.3 7239 UTR 3 XP_006712758.1
XM_011511646.2 7239 UTR 3 XP_011509948.1
XM_011511647.1 7239 UTR 3 XP_011509949.1
XM_011511648.1 7239 UTR 3 XP_011509950.1
XM_011511649.2 7239 Intron XP_011509951.1

View Full Product Details