Product Details

SNP ID
rs140217422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:174348606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCTGCTGTCATTTCTTCTGCTT[C/T]GCTCTTCACCAGGATTTCGCTGTGC
Phenotype
MIM: 605228
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CIR1 PubMed Links

Gene Details

Gene
CIR1
Gene Name
corepressor interacting with RBPJ, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004882.3 Intron NP_004873.3

View Full Product Details