Product Details

SNP ID
rs140252377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:190454513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCTAGAGAGCTTGCTTGTGTAC[A/G]ACTCTTTTTCTCTTTCTTTACCCTT
Phenotype
MIM: 613476 MIM: 616497
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MFSD6 PubMed Links

Gene Details

Gene
MFSD6
Gene Name
major facilitator superfamily domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017694.3 2263 Intron NP_060164.3
XM_005246654.2 2263 Intron XP_005246711.1
XM_005246656.3 2263 Intron XP_005246713.1
XM_011511368.1 2263 Intron XP_011509670.1
XM_017004392.1 2263 Intron XP_016859881.1
Gene
NEMP2
Gene Name
nuclear envelope integral membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142645.1 2263 Intron NP_001136117.1
XM_005246212.3 2263 Intron XP_005246269.1
XM_005246213.3 2263 Intron XP_005246270.1
XM_011510458.2 2263 UTR 3 XP_011508760.1
XM_011510459.2 2263 Intron XP_011508761.1
XM_017003098.1 2263 Intron XP_016858587.1
XM_017003099.1 2263 Intron XP_016858588.1
XM_017003100.1 2263 UTR 3 XP_016858589.1

View Full Product Details