Product Details

SNP ID
rs140378972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:101004742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGTGTTTTTTGTCACTACTGAG[A/G]GATACTGTGGGCCAGGGACTACTAA
Phenotype
MIM: 603347
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NPAS2 PubMed Links

Gene Details

Gene
NPAS2
Gene Name
neuronal PAS domain protein 2
There are no transcripts associated with this gene.

Gene
RPL31
Gene Name
ribosomal protein L31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000993.4 Intron NP_000984.1
NM_001098577.2 Intron NP_001092047.1
NM_001099693.1 Intron NP_001093163.1
Gene
TBC1D8
Gene Name
TBC1 domain family member 8
There are no transcripts associated with this gene.

View Full Product Details