Product Details
- SNP ID
-
rs140435232
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:97658066 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGAGCCCCTCACTCTCATCCCCGA[A/C]AAGGTCCGGCTGGAACAGCAGCTCG
- Phenotype
-
MIM: 605144
MIM: 123866
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ACTR1B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1042705] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACTR1B
- Gene Name
- ARP1 actin-related protein 1 homolog B, centractin beta
- Gene
- COX5B
- Gene Name
- cytochrome c oxidase subunit 5B
There are no transcripts associated with this gene.
View Full Product Details