Product Details

SNP ID
rs140530422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:169815520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTTTCCATTCTGCAGCTTTCTT[G/T]TGAACATGCTGAACATAAATAATAT
Phenotype
MIM: 109090
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
METTL5 PubMed Links

Gene Details

Gene
METTL5
Gene Name
methyltransferase like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293186.1 691 Missense Mutation CAA,CAC Q166H NP_001280115.1
NM_001293187.1 691 Missense Mutation CAA,CAC Q166H NP_001280116.1
NM_014168.3 691 Missense Mutation CAA,CAC Q166H NP_054887.2
XM_011511023.1 691 Intron XP_011509325.1
Gene
SSB
Gene Name
Sjogren syndrome antigen B
There are no transcripts associated with this gene.

View Full Product Details