Product Details

SNP ID
rs140581189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27629113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCTTCTGGGGGTCCGCGGCAC[C/T]CAGTGTGTCTGTTGGTGTTGGGAAT
Phenotype
MIM: 611479
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC121 PubMed Links

Gene Details

Gene
CCDC121
Gene Name
coiled-coil domain containing 121
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142683.2 118 Intron NP_001136155.1
NM_024584.4 118 Intron NP_078860.2
XM_005264560.3 118 Intron XP_005264617.1
Gene
GPN1
Gene Name
GPN-loop GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145047.1 118 Intron NP_001138519.1
NM_001145048.1 118 Intron NP_001138520.1
NM_001145049.1 118 Intron NP_001138521.1
NM_007266.3 118 Missense Mutation CCA,TCA P33S NP_009197.2
Gene
ZNF512
Gene Name
zinc finger protein 512
There are no transcripts associated with this gene.

View Full Product Details