Product Details

SNP ID
rs140736514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27781419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTGGTTGTGGAATCTTTAATTC[C/T]GGTTGTTTTGTGCAAATGCATGGAA
Phenotype
MIM: 610059 MIM: 611132
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL33 PubMed Links

Gene Details

Gene
MRPL33
Gene Name
mitochondrial ribosomal protein L33
There are no transcripts associated with this gene.

Gene
RBKS
Gene Name
ribokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287580.1 2335 UTR 3 NP_001274509.1
NM_022128.2 2335 UTR 3 NP_071411.1

View Full Product Details