Product Details

SNP ID
rs140962837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:61483947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCTTTTATTTACCCGTATCTGC[A/G]ACATTCCTCATAGTATGTTTGAAAG
Phenotype
MIM: 602559
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XPO1 PubMed Links

Gene Details

Gene
XPO1
Gene Name
exportin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003400.3 3395 Silent Mutation GTC,GTT V889V NP_003391.1
XM_005264544.1 3395 Silent Mutation GTC,GTT V874V XP_005264601.1
XM_005264546.1 3395 Silent Mutation GTC,GTT V758V XP_005264603.1
XM_006712094.2 3395 Silent Mutation GTC,GTT V889V XP_006712157.1
XM_011533097.1 3395 Silent Mutation GTC,GTT V889V XP_011531399.1
XM_011533098.1 3395 Silent Mutation GTC,GTT V844V XP_011531400.1
XM_011533099.2 3395 Silent Mutation GTC,GTT V823V XP_011531401.1

View Full Product Details