Product Details

SNP ID
rs141028124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:68131443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTTGACCGCTGAATAGGGTAT[G/T]CACTGAAAACAAGATCAAAATGGTA
Phenotype
MIM: 610729
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
WDR92 PubMed Links

Gene Details

Gene
WDR92
Gene Name
WD repeat domain 92
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256476.1 986 Intron NP_001243405.1
NM_138458.3 986 Missense Mutation GAA,GCA E290A NP_612467.1

View Full Product Details