Product Details
- SNP ID
-
rs141242925
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:29193498 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTTCCCTCCAGCCCCAGGTTACCC[C/T]TGTCGTGTGGCTCCTTCTTTGCTAT
- Phenotype
-
MIM: 105590
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ALK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1881421] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALK
- Gene Name
- anaplastic lymphoma receptor tyrosine kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004304.4 |
5541 |
Missense Mutation |
AAG,AGG |
K1530R |
NP_004295.2 |
- Gene
- CLIP4
- Gene Name
- CAP-Gly domain containing linker protein family member 4
There are no transcripts associated with this gene.
View Full Product Details