Product Details

SNP ID
rs141280544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:174748652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCTCGATGGCACTTTTCACCTC[C/G]GGGTGTTTGATCAGGGGAGAGTGGA
Phenotype
MIM: 100690
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CHRNA1 PubMed Links

Gene Details

Gene
CHRNA1
Gene Name
cholinergic receptor nicotinic alpha 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000079.3 1248 Silent Mutation CCC,CCG P390P NP_000070.1
NM_001039523.2 1248 Silent Mutation CCC,CCG P415P NP_001034612.1
XM_017003256.1 1248 Silent Mutation CCC,CCG P422P XP_016858745.1
XM_017003257.1 1248 Silent Mutation CCC,CCG P397P XP_016858746.1

View Full Product Details