Product Details

SNP ID
rs141393426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:109615783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCACCTACAAACTCTCACACGCC[C/T]TAGAAGATGGAGGGGACCATCACCA
Phenotype
MIM: 601181 MIM: 611737
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RANBP2 PubMed Links

Gene Details

Gene
RANBP2
Gene Name
RAN binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006267.4 1450 Intron NP_006258.3
XM_005264002.2 1450 Intron XP_005264059.1
XM_005264003.2 1450 Intron XP_005264060.1
XM_005264004.2 1450 Intron XP_005264061.1
XM_005264005.4 1450 Intron XP_005264062.1
XM_005264007.2 1450 Intron XP_005264064.1
XM_011511575.2 1450 Intron XP_011509877.1
XM_011511576.2 1450 Intron XP_011509878.1
XM_011511578.2 1450 Intron XP_011509880.1
XM_017004623.1 1450 Intron XP_016860112.1
XM_017004624.1 1450 Intron XP_016860113.1
XM_017004625.1 1450 Intron XP_016860114.1
Gene
SEPT10
Gene Name
septin 10
There are no transcripts associated with this gene.

Gene
SOWAHC
Gene Name
sosondowah ankyrin repeat domain family member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023016.3 1450 Silent Mutation CTA,TTA L432L NP_075392.2

View Full Product Details