Product Details

SNP ID
rs141509107
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:201701746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTAACAACCTAAATAACACAAAG[G/T]CCTTTCCAATTTCAACACTGTTCTT
Phenotype
MIM: 606352 MIM: 606575
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ALS2 PubMed Links

Gene Details

Gene
ALS2
Gene Name
ALS2, alsin Rho guanine nucleotide exchange factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135745.1 4727 Intron NP_001129217.1
NM_020919.3 4727 UTR 3 NP_065970.2
XM_006712654.2 4727 UTR 3 XP_006712717.1
XM_006712655.3 4727 UTR 3 XP_006712718.1
XM_011511530.2 4727 UTR 3 XP_011509832.1
XM_017004569.1 4727 UTR 3 XP_016860058.1
XM_017004570.1 4727 Intron XP_016860059.1
XM_017004571.1 4727 UTR 3 XP_016860060.1
XM_017004572.1 4727 UTR 3 XP_016860061.1
Gene
MPP4
Gene Name
membrane palmitoylated protein 4
There are no transcripts associated with this gene.

View Full Product Details