Product Details

SNP ID
rs141702314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:110128881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAAATTCAATTGATTTATCTTAC[A/G]GACCATTTTGGATTTTTGCTCTTAG
Phenotype
MIM: 607100
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NPHP1 PubMed Links

Gene Details

Gene
NPHP1
Gene Name
nephrocystin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000272.3 Intron NP_000263.2
NM_001128178.1 Intron NP_001121650.1
NM_001128179.1 Intron NP_001121651.1
NM_207181.2 Intron NP_997064.2
XM_005263676.1 Intron XP_005263733.1
XM_005263677.1 Intron XP_005263734.1
XM_005263678.2 Intron XP_005263735.1
XM_005263679.1 Intron XP_005263736.1
XM_006712551.1 Intron XP_006712614.1
XM_006712552.2 Intron XP_006712615.1
XM_011511244.1 Intron XP_011509546.1
XM_017004218.1 Intron XP_016859707.1

View Full Product Details