Product Details

SNP ID
rs141729817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:199925261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATGTTTGGTGCCCCAGAACACA[A/G]TACTGACTTTGGAGCTTTTAAGCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2orf69 PubMed Links

Gene Details

Gene
C2orf69
Gene Name
chromosome 2 open reading frame 69
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153689.5 716 Missense Mutation AAT,AGT N178S NP_710156.3
Gene
TYW5
Gene Name
tRNA-yW synthesizing protein 5
There are no transcripts associated with this gene.

View Full Product Details