Product Details

SNP ID
rs141834302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:10441657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGGTCACAGCGCTCAACAATCC[A/G]ATCGAGGCCATCACATGTTGGTCCC
Phenotype
MIM: 165640
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929715 PubMed Links

Gene Details

Gene
LOC101929715
Gene Name
uncharacterized LOC101929715
There are no transcripts associated with this gene.

Gene
ODC1
Gene Name
ornithine decarboxylase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287188.1 1654 Missense Mutation CGG,TGG R236W NP_001274117.1
NM_001287189.1 1654 Missense Mutation CGG,TGG R365W NP_001274118.1
NM_001287190.1 1654 Missense Mutation CGG,TGG R365W NP_001274119.1
NM_002539.2 1654 Missense Mutation CGG,TGG R365W NP_002530.1
Gene
SNORA80B
Gene Name
small nucleolar RNA, H/ACA box 80B
There are no transcripts associated with this gene.

View Full Product Details