Product Details

SNP ID
rs141955679
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85550518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGAAAGTTACCAAGCTTGCCAA[C/T]ATATGATGGCAATGACAAATATTGT
Phenotype
MIM: 137167 MIM: 601468
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GGCX PubMed Links

Gene Details

Gene
GGCX
Gene Name
gamma-glutamyl carboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000821.6 Intron NP_000812.2
NM_001142269.3 Intron NP_001135741.1
NM_001311312.1 Intron NP_001298241.1
XM_005264259.4 Intron XP_005264316.1
XM_011532764.2 Intron XP_011531066.1
XM_011532765.2 Intron XP_011531067.1
XM_017003803.1 Intron XP_016859292.1
Gene
MAT2A
Gene Name
methionine adenosyltransferase 2A
There are no transcripts associated with this gene.

View Full Product Details