Product Details

SNP ID
rs142007679
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:135787977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTCTTATAGGAAGGATTTTTA[C/T]GGTTTTTGCTCCCTTAACAACCCTT
Phenotype
MIM: 603202 MIM: 611216
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LCT PubMed Links

Gene Details

Gene
LCT
Gene Name
lactase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002299.3 6142 UTR 3 NP_002290.2
XM_017004088.1 6142 Intron XP_016859577.1
Gene
UBXN4
Gene Name
UBX domain protein 4
There are no transcripts associated with this gene.

View Full Product Details