Product Details

SNP ID
rs142136097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:69896536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAAAAGAAACAAAGAGCAAAGAA[C/T]GGCAGATTACTGGTAATGTTATTAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNRNP27 PubMed Links

Gene Details

Gene
SNRNP27
Gene Name
small nuclear ribonucleoprotein U4/U6.U5 subunit 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006857.2 Intron NP_006848.1

View Full Product Details