Product Details

SNP ID
rs142253999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218357701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGCATGCCTCTAGCCGAGGCTTC[C/T]TGGACAAAATGCTCTGCGGAAATTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CATIP PubMed Links

Gene Details

Gene
CATIP
Gene Name
ciliogenesis associated TTC17 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320865.1 483 Silent Mutation CTG,TTG L107L NP_001307794.1
NM_198559.1 483 Silent Mutation CTG,TTG L96L NP_940961.1
XM_005246539.4 483 Silent Mutation CTG,TTG L65L XP_005246596.1
XM_005246541.4 483 Intron XP_005246598.1
XM_011511148.2 483 Silent Mutation CTG,TTG L107L XP_011509450.1
XM_011511149.2 483 UTR 5 XP_011509451.1
XM_011511150.2 483 Intron XP_011509452.1
XM_017004053.1 483 UTR 5 XP_016859542.1
Gene
CATIP-AS1
Gene Name
CATIP antisense RNA 1
There are no transcripts associated with this gene.

Gene
CATIP-AS2
Gene Name
CATIP antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details