Product Details

SNP ID
rs142373618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27371306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGTGAATGGAGTCCTGCACTGTC[A/G]GGTGCGCTACAGCCAGCTCCTGGGG
Phenotype
MIM: 606687 MIM: 605119 MIM: 605963 MIM: 613598
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF2B4 PubMed Links

Gene Details

Gene
EIF2B4
Gene Name
eukaryotic translation initiation factor 2B subunit delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034116.1 241 Intron NP_001029288.1
NM_001318965.1 241 Intron NP_001305894.1
NM_001318966.1 241 Intron NP_001305895.1
NM_001318967.1 241 Intron NP_001305896.1
NM_001318968.1 241 Intron NP_001305897.1
NM_001318969.1 241 Intron NP_001305898.1
NM_015636.3 241 Intron NP_056451.3
NM_172195.3 241 Intron NP_751945.2
XM_006712132.1 241 Intron XP_006712195.1
XM_011533147.2 241 Intron XP_011531449.1
Gene
PPM1G
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1G
There are no transcripts associated with this gene.

Gene
SNX17
Gene Name
sorting nexin 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267059.1 241 Missense Mutation CAG,CGG Q34R NP_001253988.1
NM_001267060.1 241 Intron NP_001253989.1
NM_001267061.1 241 Missense Mutation CAG,CGG Q14R NP_001253990.1
NM_014748.3 241 Missense Mutation CAG,CGG Q34R NP_055563.1
XM_011533203.1 241 UTR 5 XP_011531505.1
XM_017005405.1 241 UTR 5 XP_016860894.1
Gene
ZNF513
Gene Name
zinc finger protein 513
There are no transcripts associated with this gene.

View Full Product Details