Product Details

SNP ID
rs142396663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:168456583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCTATCTCGCTTTTCCCCTGGC[C/T]TTCTGTATCTTCATGGTGCGGCTCA
Phenotype
MIM: 615336
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CERS6 PubMed Links

Gene Details

Gene
CERS6
Gene Name
ceramide synthase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256126.1 335 Silent Mutation GCC,GCT A45A NP_001243055.1
NM_203463.2 335 Silent Mutation GCC,GCT A45A NP_982288.1
XM_005246440.4 335 Intron XP_005246497.1
XM_017003749.1 335 Intron XP_016859238.1

View Full Product Details