Product Details

SNP ID
rs142512424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:165874814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATCCTTTCTGATTTTTGGATAAG[C/T]TGGATGTGCTTCAAGAACCTGCAAA
Phenotype
MIM: 612014
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100506124 PubMed Links

Gene Details

Gene
LOC100506124
Gene Name
uncharacterized LOC100506124
There are no transcripts associated with this gene.

Gene
TTC21B
Gene Name
tetratricopeptide repeat domain 21B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024753.4 3999 Missense Mutation ACT,GCT T1298A NP_079029.3
XM_006712761.1 3999 Intron XP_006712824.1
XM_011511871.2 3999 Missense Mutation ACT,GCT T1048A XP_011510173.1
XM_011511872.2 3999 UTR 3 XP_011510174.1
XM_017004967.1 3999 Intron XP_016860456.1
XM_017004968.1 3999 Missense Mutation ACT,GCT T1080A XP_016860457.1
XM_017004969.1 3999 Missense Mutation ACT,GCT T965A XP_016860458.1

View Full Product Details