Product Details

SNP ID
rs142565527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:203706702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCATCGTCAGGACAAAGATGCT[C/T]AGGCTGCTCTTGGCTCTCAACTTAT
Phenotype
MIM: 186760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD28 PubMed Links

Gene Details

Gene
CD28
Gene Name
CD28 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243077.1 228 Silent Mutation CTC,CTT L2L NP_001230006.1
NM_001243078.1 228 Silent Mutation CTC,CTT L2L NP_001230007.1
NM_006139.3 228 Silent Mutation CTC,CTT L2L NP_006130.1
XM_011512194.2 228 Intron XP_011510496.1
XM_011512195.2 228 Intron XP_011510497.1
XM_011512197.2 228 Silent Mutation CTC,CTT L2L XP_011510499.1

View Full Product Details