Product Details

SNP ID
rs142615527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:37171464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGAAAAGGAATGGGCCGACAGCAC[C/T]GTGTGTGTCCTGAGACTTCGCACGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SULT6B1 PubMed Links

Gene Details

Gene
SULT6B1
Gene Name
sulfotransferase family 6B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032377.1 658 Missense Mutation AGT,GGT S213G NP_001027549.1

View Full Product Details