Product Details

SNP ID
rs142616541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24793159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTTTGCGCATGCGCCCCAGCTC[C/T]TGGAGGTAAGCGGCTGTGTGCGGGT
Phenotype
MIM: 611504
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CENPO PubMed Links

Gene Details

Gene
CENPO
Gene Name
centromere protein O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199803.2 224 UTR 5 NP_001186732.1
NM_001322101.1 224 UTR 5 NP_001309030.1
NM_024322.3 224 UTR 5 NP_077298.1
Gene
PTRHD1
Gene Name
peptidyl-tRNA hydrolase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013663.1 224 Silent Mutation CAA,CAG Q73Q NP_001013685.1

View Full Product Details