Product Details

SNP ID
rs142800183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:88526428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACCCTGTGGATTTAGACATATA[C/T]CAAAGCTCCCACATGGTCGACTATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TEX37 PubMed Links

Gene Details

Gene
TEX37
Gene Name
testis expressed 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152670.2 168 Silent Mutation TAC,TAT Y17Y NP_689883.1
XM_005264188.3 168 Silent Mutation TAC,TAT Y17Y XP_005264245.3
XM_011532691.1 168 Silent Mutation TAC,TAT Y17Y XP_011530993.1
XM_011532692.1 168 Silent Mutation TAC,TAT Y17Y XP_011530994.1

View Full Product Details