Product Details

SNP ID
rs142872443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:10912880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCGTGCAGCTCATCCTGGACGAC[C/G]TGGGCGTGGACGCAGCCGAGGGCCG
Phenotype
MIM: 603787
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KCNF1 PubMed Links

Gene Details

Gene
KCNF1
Gene Name
potassium voltage-gated channel modifier subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002236.4 944 Missense Mutation CTG,GTG L152V NP_002227.2

View Full Product Details