Product Details

SNP ID
rs142904148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:114442603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGATACAGATTCCTCCAATCTGA[A/T]AGCGTTTCAGCCTCCGGAGTGAGGA
Phenotype
MIM: 608209
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DPP10 PubMed Links

Gene Details

Gene
DPP10
Gene Name
dipeptidyl peptidase like 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004360.3 246 Intron NP_001004360.2
NM_001178034.1 246 Intron NP_001171505.1
NM_001178036.1 246 Intron NP_001171507.1
NM_001178037.1 246 Intron NP_001171508.1
NM_001321905.1 246 Intron NP_001308834.1
NM_001321906.1 246 Intron NP_001308835.1
NM_001321907.1 246 UTR 5 NP_001308836.1
NM_001321908.1 246 Intron NP_001308837.1
NM_001321909.1 246 Intron NP_001308838.1
NM_001321910.1 246 UTR 5 NP_001308839.1
NM_001321911.1 246 Intron NP_001308840.1
NM_001321912.1 246 Intron NP_001308841.1
NM_001321913.1 246 UTR 5 NP_001308842.1
NM_001321914.1 246 Intron NP_001308843.1
NM_020868.4 246 UTR 5 NP_065919.2
XM_017004565.1 246 Intron XP_016860054.1
XM_017004566.1 246 Intron XP_016860055.1

View Full Product Details