Product Details

SNP ID
rs143107486
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:43837631 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATATTTTGAAATATGAATTCAC[A/G]TACATGTTAGGCAGAATTTTATACT
Phenotype
MIM: 605459 MIM: 605460
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCG5 PubMed Links

Gene Details

Gene
ABCG5
Gene Name
ATP binding cassette subfamily G member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022436.2 Intron NP_071881.1
XM_005264480.3 Intron XP_005264537.1
XM_006712073.3 Intron XP_006712136.1
XM_006712074.3 Intron XP_006712137.1
XM_011533024.2 Intron XP_011531326.1
XM_011533025.2 Intron XP_011531327.1
XM_011533026.2 Intron XP_011531328.1
XM_011533027.2 Intron XP_011531329.1
XM_011533028.2 Intron XP_011531330.1
Gene
ABCG8
Gene Name
ATP binding cassette subfamily G member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022437.2 Intron NP_071882.1
XM_005264483.3 Intron XP_005264540.1
XM_011533029.2 Intron XP_011531331.1
XM_011533030.2 Intron XP_011531332.1
Gene
DYNC2LI1
Gene Name
dynein cytoplasmic 2 light intermediate chain 1
There are no transcripts associated with this gene.

View Full Product Details